Product Details

SNP ID
rs4574070
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:23394506 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTGCCCAAGTTCAGATCAACAAAC[A/G]TGGGTGCCAGGCCTATGATGGGTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL29 PubMed Links

Gene Details

Gene
KLHL29
Gene Name
kelch like family member 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052920.1 Intron NP_443152.1
XM_006711929.3 Intron XP_006711992.1
XM_011532501.2 Intron XP_011530803.1
XM_017003264.1 Intron XP_016858753.1

View Full Product Details