Product Details

SNP ID
rs4806812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1854936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAATACAACCCTTACCCGCCCC[C/G]CGGTGGCCCAACACGAGACCCTAAT
Phenotype
MIM: 606139 MIM: 609614
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLF16 PubMed Links

Gene Details

Gene
KLF16
Gene Name
Kruppel like factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031918.3 Intron NP_114124.1
Gene
REXO1
Gene Name
RNA exonuclease 1 homolog
There are no transcripts associated with this gene.

View Full Product Details