Product Details

SNP ID
rs6075222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:17575375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCCACAGAACAACACTCTAGTCT[A/C]TGAGTAACTAGAGCTAGTCTCTTTG
Phenotype
MIM: 603307 MIM: 609114
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BFSP1 PubMed Links

Gene Details

Gene
BFSP1
Gene Name
beaded filament structural protein 1
There are no transcripts associated with this gene.

Gene
DSTN
Gene Name
destrin, actin depolymerizing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011546.1 Intron NP_001011546.1
NM_006870.3 Intron NP_006861.1
XM_011529142.1 Intron XP_011527444.1
XM_011529143.1 Intron XP_011527445.1
XM_011529144.1 Intron XP_011527446.1

View Full Product Details