Product Details
- SNP ID
-
rs35905059
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:240127158 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTAATAATTAACGGCGAATACCCA[C/G]GCAGCGCTTGGACTACTGTGCAGGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
COPS9
PubMed Links
Gene Details
- Gene
- COPS9
- Gene Name
- COP9 signalosome subunit 9
View Full Product Details