Product Details

SNP ID
rs2078616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27219036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATTTGGTTAACCTTTGTCAGTC[A/G]TCACCTTTGTCAACTCTAGACACAC
Phenotype
MIM: 114010 MIM: 604024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATRAID PubMed Links

Gene Details

Gene
ATRAID
Gene Name
all-trans retinoic acid induced differentiation factor
There are no transcripts associated with this gene.

Gene
CAD
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306079.1 Intron NP_001293008.1
NM_004341.4 Intron NP_004332.2
XM_005264555.3 Intron XP_005264612.1
XM_006712101.2 Intron XP_006712164.1
Gene
SLC5A6
Gene Name
solute carrier family 5 member 6
There are no transcripts associated with this gene.

View Full Product Details