Product Details

SNP ID
rs630418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94304801 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATCAGACTGGGTAATAAATGGG[C/T]GGGACATGGAACTCCCCTCAGCCTC
Phenotype
MIM: 615737
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IZUMO1R PubMed Links

Gene Details

Gene
IZUMO1R
Gene Name
IZUMO1 receptor, JUNO
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199206.1 Intron NP_001186135.1

View Full Product Details