Product Details

SNP ID
rs1320644
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:76659143 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCACAGCCGGGGCAGGACTCACC[A/G]CACCAAACCAGCGGGGCCTGCCGAG
Phenotype
MIM: 137207
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC32 PubMed Links

Gene Details

Gene
LRRC32
Gene Name
leucine rich repeat containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128922.1 2675 UTR 3 NP_001122394.1
NM_005512.2 2675 UTR 3 NP_005503.1
XM_005273902.3 2675 Intron XP_005273959.1
XM_005273903.4 2675 UTR 3 XP_005273960.2
XM_011544935.2 2675 UTR 3 XP_011543237.1
XM_017017527.1 2675 Intron XP_016873016.1

View Full Product Details