Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015566.2 | 6027 | UTR 3 | NP_056381.1 | ||
XM_005248480.2 | 6027 | UTR 3 | XP_005248537.1 | ||
XM_011543306.2 | 6027 | UTR 3 | XP_011541608.1 | ||
XM_011543307.2 | 6027 | UTR 3 | XP_011541609.1 | ||
XM_017009332.1 | 6027 | UTR 3 | XP_016864821.1 | ||
XM_017009333.1 | 6027 | UTR 3 | XP_016864822.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271665.1 | 6027 | Intron | NP_001258594.1 | ||
NM_014886.4 | 6027 | Intron | NP_055701.1 | ||
XM_011543098.1 | 6027 | Intron | XP_011541400.1 | ||
XM_017008952.1 | 6027 | Intron | XP_016864441.1 | ||
XM_017008953.1 | 6027 | Intron | XP_016864442.1 |