Product Details

SNP ID
hCV2142049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:135809909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCCTCTCTACAAGCCAAAAGTC[A/G]CAAGGCTAATATTTACTCCCCTGAT
Phenotype
MIM: 604645
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928326 PubMed Links

Gene Details

Gene
LOC101928326
Gene Name
uncharacterized LOC101928326
There are no transcripts associated with this gene.

Gene
PDE7B
Gene Name
phosphodiesterase 7B
There are no transcripts associated with this gene.

View Full Product Details