Product Details

SNP ID
rs11033046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35253386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATCTCTAGCACTCATCCTCAG[A/T]CTCTGCTACTCAAAGTTTTCCTAAG
Phenotype
MIM: 600300
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC1A2 PubMed Links

Gene Details

Gene
SLC1A2
Gene Name
solute carrier family 1 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195728.2 9825 UTR 3 NP_001182657.1
NM_001252652.1 9825 UTR 3 NP_001239581.1
NM_004171.3 9825 UTR 3 NP_004162.2
XM_005253067.1 9825 UTR 3 XP_005253124.1
XM_011520285.2 9825 UTR 3 XP_011518587.1
XM_017018136.1 9825 UTR 3 XP_016873625.1
XM_017018137.1 9825 UTR 3 XP_016873626.1
XM_017018138.1 9825 UTR 3 XP_016873627.1
XM_017018139.1 9825 UTR 3 XP_016873628.1

View Full Product Details