Product Details

SNP ID
rs35134599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46917031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTCCGGACAGGAAAGACAAAGA[A/C]AGTGGCTAAGTAGTTAGATGCCTTG
Phenotype
MIM: 607788 MIM: 609332
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MCFD2 PubMed Links

Gene Details

Gene
MCFD2
Gene Name
multiple coagulation factor deficiency 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171506.2 Intron NP_001164977.1
NM_001171507.2 Intron NP_001164978.1
NM_001171508.2 Intron NP_001164979.1
NM_001171509.2 Intron NP_001164980.1
NM_001171510.2 Intron NP_001164981.1
NM_001171511.2 Intron NP_001164982.1
NM_139279.5 Intron NP_644808.1
Gene
TTC7A
Gene Name
tetratricopeptide repeat domain 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288951.1 Intron NP_001275880.1
NM_001288953.1 Intron NP_001275882.1
NM_001288955.1 Intron NP_001275884.1
NM_020458.3 Intron NP_065191.2
XM_005264439.3 Intron XP_005264496.1
XM_011532998.2 Intron XP_011531300.1
XM_011532999.2 Intron XP_011531301.1
XM_011533000.2 Intron XP_011531302.1
XM_011533001.2 Intron XP_011531303.1
XM_017004524.1 Intron XP_016860013.1
XM_017004525.1 Intron XP_016860014.1
XM_017004526.1 Intron XP_016860015.1
XM_017004527.1 Intron XP_016860016.1
XM_017004528.1 Intron XP_016860017.1
XM_017004529.1 Intron XP_016860018.1
XM_017004530.1 Intron XP_016860019.1

View Full Product Details