Product Details
- SNP ID
-
rs12622481
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:238065212 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTCCTATCCATTTGCTTATTAAA[C/T]GTTTCACGGCAACTTAATAACAAAG
- Phenotype
-
MIM: 611056
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SCLY
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80221317] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCLY
- Gene Name
- selenocysteine lyase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016510.5 |
|
Intron |
|
|
NP_057594.4 |
- Gene
- UBE2F-SCLY
- Gene Name
- UBE2F-SCLY readthrough (NMD candidate)
There are no transcripts associated with this gene.
View Full Product Details