Product Details
- SNP ID
-
rs11691216
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:238139455 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGTGTACCGGCCGGGATGCTGGGG[C/T]GTGGGGCCCCGGCCACGTGAGCTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ESPNL
PubMed Links
Gene Details
- Gene
- ESPNL
- Gene Name
- espin-like
There are no transcripts associated with this gene.
- Gene
- KLHL30
- Gene Name
- kelch like family member 30
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198582.3 |
|
Intron |
|
|
NP_940984.3 |
View Full Product Details