Product Details

SNP ID
rs35047785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238162603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGGAGAAGCTTTCACTGAGGT[A/G]GATCCTTGGCTGGAGGTTGGAGCCC
Phenotype
MIM: 615099
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERFE PubMed Links

Gene Details

Gene
ERFE
Gene Name
erythroferrone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291832.1 Intron NP_001278761.1
Gene
ILKAP
Gene Name
ILK associated serine/threonine phosphatase
There are no transcripts associated with this gene.

Gene
KLHL30
Gene Name
kelch like family member 30
There are no transcripts associated with this gene.

View Full Product Details