Product Details

SNP ID
rs503156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66053202 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTCACGTGCATGTGTGAGATGTG[A/G]CACCCTTGCACATTACTCGCCTGAC
Phenotype
MIM: 608234 MIM: 605591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAL3ST3 PubMed Links

Gene Details

Gene
GAL3ST3
Gene Name
galactose-3-O-sulfotransferase 3
There are no transcripts associated with this gene.

Gene
SF3B2
Gene Name
splicing factor 3b subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006842.2 Intron NP_006833.2
XM_005273726.3 Intron XP_005273783.1
XM_011544740.2 Intron XP_011543042.1
XM_017017144.1 Intron XP_016872633.1

View Full Product Details