Product Details

SNP ID
rs12603194
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:74758785 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCCAGCCCTGCCCCAGCTTGGCGGT[A/G]AGGGTCCAAGAGCAATGGTGGCTGA
Phenotype
MIM: 604990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC9A3R1 PubMed Links

Gene Details

Gene
SLC9A3R1
Gene Name
SLC9A3 regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004252.4 Intron NP_004243.1

View Full Product Details