Product Details

SNP ID
rs1466113
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:73169179 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAGAAATGTGTAAATTTGGTGAGA[C/G]TTTAAACAGCCTGTGACCGACGGGC
Phenotype
MIM: 182452
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SSTR2 PubMed Links

Gene Details

Gene
SSTR2
Gene Name
somatostatin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001050.2 Intron NP_001041.1

View Full Product Details