Product Details

SNP ID
rs9652403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103569687 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCCACAGTCTAGGGGGGTGGACC[C/G]TGGTGCCGTCCCTTTGGTGGGACTC
Phenotype
MIM: 616003 MIM: 603885
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOPT1 PubMed Links

Gene Details

Gene
APOPT1
Gene Name
apoptogenic 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302652.1 Intron NP_001289581.1
NM_001302653.1 Intron NP_001289582.1
NM_001302654.1 Intron NP_001289583.1
NM_032374.4 Intron NP_115750.2
Gene
BAG5
Gene Name
BCL2 associated athanogene 5
There are no transcripts associated with this gene.

View Full Product Details