Product Details

SNP ID
rs11904706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:12717493 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACCTCCGCGCGGCGGGCCGAGCC[C/T]AGGGCTTTGTCGCGGTACCTGCGCC
Phenotype
MIM: 609462
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIB2 PubMed Links

Gene Details

Gene
TRIB2
Gene Name
tribbles pseudokinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021643.3 622 UTR 5 NP_067675.1

View Full Product Details