Product Details

SNP ID
rs11765965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66377234 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCCGGGTCTCTCAGGGCCGCCGG[A/G]AAGCCGAAGGTGGGTCCGGGAGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00174 PubMed Links

Gene Details

Gene
LINC00174
Gene Name
long intergenic non-protein coding RNA 174
There are no transcripts associated with this gene.

View Full Product Details