Product Details
- SNP ID
-
rs10883887
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:103597424 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTGCTGGGAGTCATTTTTAGGCCA[A/G]GTCTCTCTGAGCAGCTGGCTGCACC
- Phenotype
-
MIM: 603804
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NEURL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34313279] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NEURL1
- Gene Name
- neuralized E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.
- Gene
- SH3PXD2A
- Gene Name
- SH3 and PX domains 2A
View Full Product Details