Product Details

SNP ID
rs175512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75056679 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCAACACAGAAAAAGAATTGTA[C/T]ACCATGACAAACTAGGATTTCTTTC
Phenotype
MIM: 600875 MIM: 604395
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACYP1 PubMed Links

Gene Details

Gene
ACYP1
Gene Name
acylphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001107.4 Intron NP_001098.1
NM_001302616.1 Intron NP_001289545.1
NM_001302617.1 Intron NP_001289546.1
Gene
MLH3
Gene Name
mutL homolog 3
There are no transcripts associated with this gene.

View Full Product Details