Product Details

SNP ID
rs12168831
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:25749688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGCATGCCACGTGGATGTCAGGG[C/T]GGTCGTCTTGTTCCCCATTACTCAA
Phenotype
MIM: 607295
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MYO18B PubMed Links
Additional Information
For this assay, SNP(s) [rs114989234] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MYO18B
Gene Name
myosin XVIIIB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318245.1 Intron NP_001305174.1
NM_032608.6 Intron NP_115997.5
XM_011530458.2 Intron XP_011528760.1
XM_011530459.2 Intron XP_011528761.1
XM_011530460.2 Intron XP_011528762.1
XM_011530461.2 Intron XP_011528763.1
XM_011530464.2 Intron XP_011528766.1
XM_011530465.2 Intron XP_011528767.1
XM_011530466.2 Intron XP_011528768.1
XM_017029012.1 Intron XP_016884501.1
XM_017029013.1 Intron XP_016884502.1
XM_017029014.1 Intron XP_016884503.1
XM_017029015.1 Intron XP_016884504.1
XM_017029016.1 Intron XP_016884505.1
XM_017029017.1 Intron XP_016884506.1

View Full Product Details