Product Details

SNP ID
rs12133334
Assay Type
Functionally tested
NCBI dbSNP Submissions
62
Location
Chr.1:17067879 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTGGCCCTAAAGGAACTTTTAA[A/C]GATTGAAACTGAGTCTTTTCAGTTG
Phenotype
MIM: 607935
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PADI2 PubMed Links

Gene Details

Gene
PADI2
Gene Name
peptidyl arginine deiminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007365.2 2419 UTR 3 NP_031391.2
XM_017000148.1 2419 UTR 3 XP_016855637.1

View Full Product Details