Product Details

SNP ID
rs2746473
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:17023224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCTGAGATCAGCAGGCTGAAA[C/T]GAGTTAATCACATGACCATAGGCCT
Phenotype
MIM: 185470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDHB PubMed Links

Gene Details

Gene
SDHB
Gene Name
succinate dehydrogenase complex iron sulfur subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003000.2 Intron NP_002991.2

View Full Product Details