Product Details

SNP ID
rs761420
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:17022352 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCAGGTACCTGCTCATGGGTTCCT[C/G]TGCATCCTTAACTCCCACAATGTCC
Phenotype
MIM: 185470
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SDHB PubMed Links

Gene Details

Gene
SDHB
Gene Name
succinate dehydrogenase complex iron sulfur subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003000.2 Intron NP_002991.2

View Full Product Details