Product Details

SNP ID
rs7790757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107898058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTAATTTTAATAACATTAATTGC[C/G]TTGTTTAAGTCAGTCACCTCTTATT
Phenotype
MIM: 238331
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DLD PubMed Links

Gene Details

Gene
DLD
Gene Name
dihydrolipoamide dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000108.4 Intron NP_000099.2
NM_001289750.1 Intron NP_001276679.1
NM_001289751.1 Intron NP_001276680.1
NM_001289752.1 Intron NP_001276681.1

View Full Product Details