Product Details

SNP ID
rs6707144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:10146699 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTGGGGATGGATCAGGCCTTGAG[A/G]GGGTGCCTGGTAAGCCCCGGGAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf48 PubMed Links

Gene Details

Gene
C2orf48
Gene Name
chromosome 2 open reading frame 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182626.3 Intron NP_872432.1

View Full Product Details