Product Details

SNP ID
rs165598
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:20862407 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GAGGGATGTTCTTAGGGAAGACAGG[A/C]AAATATGCAAATGCACTGGCCCAGC
Phenotype
MIM: 600286 MIM: 604202
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PI4KA PubMed Links

Gene Details

Gene
PI4KA
Gene Name
phosphatidylinositol 4-kinase alpha
There are no transcripts associated with this gene.

Gene
SNAP29
Gene Name
synaptosome associated protein 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004782.3 Intron NP_004773.1

View Full Product Details