Product Details
- SNP ID
-
rs118121
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:116155168 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCTCCACGCCATCCCATGTCCTA[G/T]CTTTTAAAGCCCATTGCCTGTCACT
- Phenotype
-
MIM: 176385
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PAPPA
PubMed Links
Gene Details
- Gene
- PAPPA
- Gene Name
- pappalysin 1
View Full Product Details