Product Details

SNP ID
rs131476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24721953 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATGAACAACAGAGCTGACTATA[C/T]GAAACTATTTTGTAGCATATTCAGT
Phenotype
MIM: 610314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIWIL3 PubMed Links

Gene Details

Gene
PIWIL3
Gene Name
piwi like RNA-mediated gene silencing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008496.3 Intron NP_001008496.2
NM_001255975.1 Intron NP_001242904.1

View Full Product Details