Product Details

SNP ID
rs140502
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:23264357 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCACGTCCTGGATATTCAAAACCCG[C/T]GACCGTCAGGGCCACCCCTGCCTGT
Phenotype
MIM: 151410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCR PubMed Links

Gene Details

Gene
BCR
Gene Name
BCR, RhoGEF and GTPase activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004327.3 Intron NP_004318.3
NM_021574.2 Intron NP_067585.2
Gene
FBXW4P1
Gene Name
F-box and WD repeat domain containing 4 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details