Product Details
- SNP ID
-
rs121908738
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44622612 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAATGACTGCATGCTCCGTGTCC[A/G]GCTTCCAGGCACCAGTGAGGTAGCT
- Phenotype
-
MIM: 608958
MIM: 604932
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116962828] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADA
- Gene Name
- adenosine deaminase
- Gene
- PKIG
- Gene Name
- protein kinase (cAMP-dependent, catalytic) inhibitor gamma
There are no transcripts associated with this gene.
View Full Product Details