Product Details

SNP ID
rs203862
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:167925347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGTAGAATAATAAATAAAAAGG[A/G]AAGTTAGCCAGTAGAGTTGGAAGCA
Phenotype
MIM: 605205 MIM: 614737
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY10 PubMed Links

Gene Details

Gene
ADCY10
Gene Name
adenylate cyclase 10, soluble
There are no transcripts associated with this gene.

Gene
MPC2
Gene Name
mitochondrial pyruvate carrier 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143674.3 Intron NP_001137146.1
NM_015415.3 Intron NP_056230.1
XM_006711266.3 Intron XP_006711329.1

View Full Product Details