Product Details
- SNP ID
-
rs109413
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:29307156 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGGGGCCGCGGGATGAGCCAGGT[G/T]AGCGCGGAGACCCCCAGCACTCCCT
- Phenotype
-
MIM: 133450
MIM: 602128
MIM: 602220
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EWSR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs376400273] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EWSR1
- Gene Name
- EWS RNA binding protein 1
There are no transcripts associated with this gene.
- Gene
- GAS2L1
- Gene Name
- growth arrest specific 2 like 1
- Gene
- RASL10A
- Gene Name
- RAS like family 10 member A
There are no transcripts associated with this gene.
View Full Product Details