Product Details
- SNP ID
-
rs989735224
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:150577357 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCATTGCTGGGCCATATGTTTAGT[G/T]TATGTTTAACTTTACAAGGAACTGT
- Phenotype
-
MIM: 300415
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MTM1
PubMed Links
Gene Details
- Gene
- MTM1
- Gene Name
- myotubularin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000252.2 |
|
Intron |
|
|
NP_000243.1 |
View Full Product Details