Product Details

SNP ID
rs135623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44494792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTAGGGCTTTCAAAAAGCCACT[C/T]GAGTAGAGTGAGCGCTGGCGACACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDOC1L PubMed Links

Gene Details

Gene
LDOC1L
Gene Name
leucine zipper down-regulated in cancer 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032287.2 3102 UTR 3 NP_115663.2

View Full Product Details