Product Details

SNP ID
rs215212
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:37493898 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTCCTGCAGTTCTGTTACTAAAAAT[C/G]ACATAAAGTAAGACCCAATGTCTTA
Phenotype
MIM: 611001 MIM: 610562
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MEAF6 PubMed Links

Gene Details

Gene
MEAF6
Gene Name
MYST/Esa1 associated factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270875.1 880 UTR 3 NP_001257804.1
NM_001270876.1 880 Intron NP_001257805.1
NM_022756.5 880 UTR 3 NP_073593.2
Gene
MIR5581
Gene Name
microRNA 5581
There are no transcripts associated with this gene.

Gene
ZC3H12A
Gene Name
zinc finger CCCH-type containing 12A
There are no transcripts associated with this gene.

View Full Product Details