Product Details
- SNP ID
-
rs195860
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:98835500 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCGCCGGCGGGCAGCCGGCCGG[G/T]CTGCACCACCACGGCCTGCGGGACG
- Phenotype
-
MIM: 600494
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
POU3F2
PubMed Links
Gene Details
- Gene
- POU3F2
- Gene Name
- POU class 3 homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005604.3 |
797 |
Silent Mutation |
GGG,GGT |
G209G |
NP_005595.2 |
View Full Product Details