Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001324191.1 | 2441 | Intron | NP_001311120.1 | ||
NM_001324192.1 | 2441 | Intron | NP_001311121.1 | ||
NM_001324193.1 | 2441 | Intron | NP_001311122.1 | ||
NM_024960.5 | 2441 | Intron | NP_079236.3 | ||
NM_153638.3 | 2441 | Intron | NP_705902.2 | ||
NM_153640.3 | 2441 | Intron | NP_705904.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001134337.2 | 2441 | UTR 3 | NP_001127809.1 | ||
NM_001134338.2 | 2441 | UTR 3 | NP_001127810.1 | ||
NM_001321749.1 | 2441 | UTR 3 | NP_001308678.1 | ||
NM_007219.4 | 2441 | UTR 3 | NP_009150.1 | ||
XM_011529145.2 | 2441 | UTR 3 | XP_011527447.1 | ||
XM_011529146.2 | 2441 | UTR 3 | XP_011527448.1 | ||
XM_011529147.2 | 2441 | Intron | XP_011527449.1 | ||
XM_017027620.1 | 2441 | UTR 3 | XP_016883109.1 | ||
XM_017027621.1 | 2441 | UTR 3 | XP_016883110.1 | ||
XM_017027622.1 | 2441 | Intron | XP_016883111.1 |