Product Details

SNP ID
rs745515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24962592 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCATCTGTAAGCTGGGAGTAATAA[G/T]AGAGCTGGCCTCATGAGGTTGACAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1671 PubMed Links

Gene Details

Gene
KIAA1671
Gene Name
KIAA1671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145206.1 Intron NP_001138678.1
XM_005261793.2 Intron XP_005261850.1
XM_005261794.4 Intron XP_005261851.1
XM_006724346.2 Intron XP_006724409.1

View Full Product Details