Product Details
- SNP ID
-
rs739442
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:132894747 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTCTCTCTGCTCGAGGCCTCCGT[A/G]GGCACTAAGATTTCGTACCGTGACA
- Phenotype
-
MIM: 605284
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPACA9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79277527] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPACA9
- Gene Name
- sperm acrosome associated 9
There are no transcripts associated with this gene.
- Gene
- TSC1
- Gene Name
- tuberous sclerosis 1
View Full Product Details