Product Details

SNP ID
rs378740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:134025754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGGCATCCCAGGAGGGGAGGTC[A/C]CTGACGCTGCGCAGCTGGGAAGGCG
Phenotype
MIM: 601541
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BRD3 PubMed Links

Gene Details

Gene
BRD3
Gene Name
bromodomain containing 3
There are no transcripts associated with this gene.

Gene
LINC00094
Gene Name
long intergenic non-protein coding RNA 94
There are no transcripts associated with this gene.

View Full Product Details