Product Details

SNP ID
rs406446
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:10711125 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATAGAATTATGAATTACAAGAAAG[A/G]AAGAGCCTTCCATTGTTAGTCTAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADPRM PubMed Links

Gene Details

Gene
ADPRM
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020233.4 2169 Missense Mutation GAA,GGA E337G NP_064618.3
XM_005256738.2 2169 UTR 3 XP_005256795.1
Gene
MAGOH2P
Gene Name
mago homolog 2, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM220
Gene Name
transmembrane protein 220
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004313.1 2169 Intron NP_001004313.1
XM_005256634.4 2169 Intron XP_005256691.1
XM_011523835.2 2169 Intron XP_011522137.1
XM_011523836.2 2169 Intron XP_011522138.1
XM_017024616.1 2169 UTR 3 XP_016880105.1
XM_017024617.1 2169 UTR 3 XP_016880106.1

View Full Product Details