Product Details
- SNP ID
-
hCV2257352
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:155802982 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGCCTATTTGGTAGAGCAGCTGC[A/G]AGTACCAGTGGATGCCCGCGGTGGC
- Phenotype
-
MIM: 600725
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SHH
PubMed Links
Gene Details
- Gene
- SHH
- Gene Name
- sonic hedgehog
View Full Product Details