Product Details

SNP ID
rs14304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36071135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAATGGCCCTGCCACCCTGGAGGC[C/T]ACCTCTTCTAAGAGTCCCATCTGCT
Phenotype
MIM: 603757
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL18 PubMed Links

Gene Details

Gene
CCL18
Gene Name
C-C motif chemokine ligand 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002988.3 435 UTR 3 NP_002979.1

View Full Product Details