Product Details

SNP ID
rs421313
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119470226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGTACCAACCTAACAGTCCAAG[A/C]GCTAAAGAGTTGTTTTTTTAATTGC
Phenotype
MIM: 300150
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
SLC25A5 PubMed Links

Gene Details

Gene
SLC25A5
Gene Name
solute carrier family 25 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001152.4 Intron NP_001143.2
Gene
SLC25A5-AS1
Gene Name
SLC25A5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details