Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002214.2 | Intron | NP_002205.1 | |||
XM_011515393.2 | Intron | XP_011513695.1 | |||
XM_011515394.2 | Intron | XP_011513696.1 | |||
XM_011515396.1 | Intron | XP_011513698.1 | |||
XM_017012178.1 | Intron | XP_016867667.1 | |||
XM_017012179.1 | Intron | XP_016867668.1 | |||
XM_017012180.1 | Intron | XP_016867669.1 | |||
XM_017012181.1 | Intron | XP_016867670.1 | |||
XM_017012182.1 | Intron | XP_016867671.1 | |||
XM_017012183.1 | Intron | XP_016867672.1 |