Product Details

SNP ID
rs739993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17743723 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCACAGCTCCTGCGGCTCCCTT[C/G]CGCCAGCCTCCTCCCTGGTCCTCAT
Phenotype
MIM: 176258
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNC1 PubMed Links

Gene Details

Gene
KCNC1
Gene Name
potassium voltage-gated channel subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112741.1 Intron NP_001106212.1
NM_004976.4 Intron NP_004967.1
XM_011520080.2 Intron XP_011518382.1

View Full Product Details