Product Details

SNP ID
rs38409
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:30499035 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATAATAGTCAAGATGGTGTATGGGA[C/T]TAAACATTTAACTACATCTGAACAT
Phenotype
MIM: 137170
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGCT PubMed Links

Gene Details

Gene
GGCT
Gene Name
gamma-glutamylcyclotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199815.1 Intron NP_001186744.1
NM_001199816.1 Intron NP_001186745.1
NM_001199817.1 Intron NP_001186746.1
NM_024051.3 Intron NP_076956.1

View Full Product Details